Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IRF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002199.3 | NP_002190.2 | 3, 4 | 395 | ||
GenBank mRNA | AK312953.1 | - | 3, 4 | 373 | ||
BC015803.2 | - | 2, 3 | 202 | 3920890 | ||
BP200998.1 | - | 3, 4 | 395 | |||
BT007264.1 | - | 2, 3 | 195 | |||
CR457077.1 | - | 2, 3 | 195 | |||
X15949.1 | - | 3, 4 | 293 |