Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MT1H | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005951.2 | NP_005942.1 | 3 | 276 | ||
GenBank mRNA | BC008408.1 | - | 3 | 276 | 4274128 | |
BC059780.1 | - | 3 | 256 | 5227592 | ||
BC146930.1 | - | 3 | 227 | 9056858 | ||
BC146934.1 | - | 3 | 227 | 9056862 | ||
X64177.1 | - | 3 | 228 |