Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AFF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025108.1 | NP_001020279.1 | 4 | 313 | ||
NM_002285.2 | NP_002276.2 | 4 | 330 | |||
XM_005263943.3 | XP_005264000.2 | 3 | 207 | |||
XM_011511169.2 | XP_011509471.1 | 4 | 313 | |||
XM_011511170.2 | XP_011509472.1 | 5 | 449 | |||
XM_011511171.2 | XP_011509473.1 | 4 | 313 | |||
XM_011511172.2 | XP_011509474.1 | 4 | 313 | |||
XM_011511173.2 | XP_011509475.1 | 4 | 326 | |||
XM_011511174.2 | XP_011509476.1 | 4 | 270 | |||
XM_011511175.2 | XP_011509477.1 | 6 | 560 | |||
XM_011511176.2 | XP_011509478.1 | 3 | 251 | |||
XM_011511177.2 | XP_011509479.1 | 4 | 341 | |||
GenBank mRNA | AK092327.1 | - | 5 | 403 | ||
AK290468.1 | - | 3 | 188 | |||
AK297407.1 | - | 5 | 581 | |||
AK308645.1 | - | 5 | 449 | |||
BC036895.1 | - | 4 | 275 | |||
BC136579.1 | - | 2 | 113 | 9020569 | ||
BC144266.1 | - | 3 | 188 | 9052786 | ||
BI906088.1 | - | 4 | 329 | |||
U34360.1 | - | 2 | 148 |