Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPSA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304288.1 | NP_001291217.1 | 2, 3 | 239 | ||
NM_002295.5 | NP_002286.2 | 2, 3 | 239 | |||
GenBank mRNA | AF503367.1 | - | 1 | 199 | ||
BC005391.1 | - | 1 | 211 | 3997019 | ||
BC008867.2 | - | 1 | 160 | 4130936 | ||
BC010418.2 | - | 1 | 191 | 4079845 | ||
BC013827.2 | - | 1 | 195 | 3446816 | ||
BC018867.2 | - | 1 | 185 | |||
BC034537.1 | - | 1 | 192 | 3455545 | ||
BC050688.1 | - | 1 | 194 | 4443949 | ||
BC053370.1 | - | 1 | 192 | 5108337 | ||
BC062714.1 | - | 1 | 215 | 3686924 | ||
BC066941.1 | - | 2, 3 | 208 | 4820432 | ||
BC068062.1 | - | 1 | 173 | 5427114 | ||
BC070263.1 | - | 2, 3 | 239 | 3996864 | ||
BC071693.1 | - | 1 | 201 | 3884457 | ||
BC071968.1 | - | 1 | 192 | 5814758 | ||
BC071969.1 | - | 1 | 180 | 6483856 | ||
BC071970.1 | - | 1 | 165 | 6484091 | ||
BC073863.1 | - | 1 | 177 | 3924066 | ||
BC107567.1 | - | 1 | 212 | 3681016 | ||
BT007219.1 | - | 1, 2 | 130 | |||
BU932677.1 | - | 169 | ||||
J03799.1 | - | 1, 2 | 167 | |||
S37431.1 | - | 1, 2 | 145 | |||
X15005.1 | - | 2, 3 | 193 | |||
X61156.1 | - | 1, 2 | 100 |
Gene Symbol: RPSAP15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF284768.1 | - | 1 | 207 |
Gene Symbol: RPSAP19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BX648992.1 | - | 1 | 1709 |
Gene Symbol: RPSAP58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003662.3 | 4 | 553 |