Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHIC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012110.3 | NP_036242.1 | 2 | 487 | ||
XM_006714037.3 | XP_006714100.1 | 2 | 487 | |||
XM_011534382.2 | XP_011532684.1 | 2 | 487 | |||
GenBank mRNA | AF159423.1 | - | 2 | 388 | ||
AK312427.1 | - | 2 | 450 | |||
BC034691.1 | - | 2 | 399 | 4747392 | ||
BI828538.1 | - | 2 | 460 |