Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PREB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013388.4 | NP_037520.1 | 9 | 1801 | ||
XM_006711914.2 | XP_006711977.1 | 8 | 1608 | |||
XM_011532471.2 | XP_011530773.1 | 8 | 1622 | |||
XM_011532472.2 | XP_011530774.1 | 7 | 1448 | |||
GenBank mRNA | AF203687.1 | - | 9 | 1679 | ||
AF226684.1 | - | 9 | 1692 | |||
AK001976.1 | - | 3 | 1874 | |||
AK023064.1 | - | 9 | 1758 | |||
BC002765.2 | - | 9 | 1732 | 3631721 | ||
BC012890.1 | - | 9 | 1742 | 4158577 | ||
BC016472.1 | - | 9 | 1732 | 4178911 | ||
BC016906.1 | - | 9 | 1675 | 4427483 | ||
BC041032.1 | - | 9 | 1795 | 4638313 |