Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PREB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013388.4 | NP_037520.1 | 9 | 1803 | ||
XM_006711914.2 | XP_006711977.1 | 8 | 1610 | |||
XM_011532471.2 | XP_011530773.1 | 8 | 1624 | |||
XM_011532472.2 | XP_011530774.1 | 7 | 1450 | |||
GenBank mRNA | AF203687.1 | - | 9 | 1681 | ||
AF226684.1 | - | 9 | 1694 | |||
AK001976.1 | - | 3 | 1876 | |||
AK023064.1 | - | 9 | 1760 | |||
BC002765.2 | - | 9 | 1734 | 3631721 | ||
BC012890.1 | - | 9 | 1744 | 4158577 | ||
BC016472.1 | - | 9 | 1734 | 4178911 | ||
BC016906.1 | - | 9 | 1677 | 4427483 | ||
BC041032.1 | - | 9 | 1797 | 4638313 |