Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321241.1 | NP_001308170.1 | 4 | 356 | ||
NM_016016.3 | NP_057100.1 | 4 | 356 | |||
XM_011524884.1 | XP_011523186.1 | 4 | 331 | |||
XM_017024737.1 | XP_016880226.1 | 4 | 331 | |||
GenBank mRNA | AB209576.1 | - | 3 | 766 | ||
AF119864.1 | - | 3 | 142 | |||
AF151827.1 | - | 4 | 298 | |||
AK026060.1 | - | 4 | 298 | |||
AK295478.1 | - | 4 | 317 | |||
AL133584.1 | - | 4 | 298 | |||
BC001398.2 | - | 4 | 302 | 3051436 | ||
BC009330.2 | - | 4 | 274 | 4126268 | ||
CB960224.1 | - | 4 | 383 | |||
CV573041.1 | - | 4 | 335 |