Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FKBP11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143782.1 | NP_001137254.1 | 2 | 299 | ||
NM_016594.2 | NP_057678.1 | 2 | 299 | |||
GenBank mRNA | AF238079.1 | - | 2 | 252 | ||
AK301455.1 | - | 2 | 272 | |||
AY358998.1 | - | 2 | 257 | |||
BC027973.1 | - | 2 | 251 | 5229643 | ||
BC048292.1 | - | 2 | 262 | 5207367 | ||
DA378205.1 | - | 2 | 299 |