Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OR3A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002551.3 | NP_002542.3 | 1 | 696 | ||
XM_017024697.1 | XP_016880186.1 | 6 | 1316 | |||
XM_017024698.1 | XP_016880187.1 | 4 | 1008 | |||
GenBank mRNA | BC095517.1 | - | 1 | 696 | 6971961 |