Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATAD3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170535.1 | NP_001164006.1 | 10 | 1162 | ||
NM_001170536.1 | NP_001164007.1 | 10 | 992 | |||
NM_018188.3 | NP_060658.3 | 10 | 1306 | |||
XM_017001628.1 | XP_016857117.1 | 10 | 1162 | |||
GenBank mRNA | AK001571.1 | - | 10 | 1145 | ||
AK025865.1 | - | 3 | 162 | |||
AK056099.1 | - | 10 | 992 | |||
AK091144.1 | - | 10 | 1274 | |||
AK310572.1 | - | 1184 | ||||
BC007803.2 | - | 10 | 1123 | 4136170 | ||
BC011814.2 | - | 10 | 1128 | 3162989 | ||
BC014101.2 | - | 10 | 1152 | 3503438 | ||
BC033109.1 | - | 10 | 1126 | 4877449 | ||
BC063607.1 | - | 10 | 1120 | 4939339 | ||
GU189416.1 | - | 10 | 1066 |