Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MCOLN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020533.2 | NP_065394.1 | 2 | 357 | ||
GenBank mRNA | AF249319.1 | - | 2 | 300 | ||
AF287269.1 | - | 2 | 341 | |||
AJ293659.1 | - | 2 | 296 | |||
AJ293970.1 | - | 2 | 296 | |||
AK026102.1 | - | 2 | 311 | |||
AK222673.1 | - | 2 | 336 | |||
AK294330.1 | - | 2 | 322 | |||
AK310109.1 | - | 2 | 332 | |||
BC005149.2 | - | 2 | 339 | 3507836 | ||
DC356874.1 | - | 2 | 360 |