Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RETN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193374.1 | NP_001180303.1 | 2 | 124 | ||
NM_020415.3 | NP_065148.1 | 2 | 133 | |||
GenBank mRNA | AB111910.1 | - | 1 | 87 | ||
AF205952.1 | - | 1 | 111 | |||
AF290874.1 | - | 2 | 133 | |||
AF323081.1 | - | 2 | 133 | |||
AY207314.1 | - | 2 | 124 | |||
AY359066.1 | - | 1 | 111 | |||
BC069302.1 | - | 2 | 133 | 7262685 | ||
BC101554.1 | - | 2 | 116 | 8069060 | ||
BC101560.1 | - | 2 | 116 | 8069066 |