Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSKS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021733.1 | NP_068379.1 | 2 | 401 | ||
XM_011527201.1 | XP_011525503.1 | 2 | 471 | |||
XM_011527202.1 | XP_011525504.1 | 2 | 471 | |||
XM_011527203.2 | XP_011525505.1 | 2 | 471 | |||
GenBank mRNA | BC058862.1 | - | 2 | 429 | 5170446 |