Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143780.2 | NP_001137252.1 | 12 | 1491 | ||
NM_001321240.1 | NP_001308169.1 | 11 | 1422 | |||
NM_001321241.1 | NP_001308170.1 | 12 | 1485 | |||
NM_016016.3 | NP_057100.1 | 12 | 1467 | |||
XM_011524880.2 | XP_011523182.1 | 12 | 1466 | |||
XM_011524882.2 | XP_011523184.1 | 11 | 1406 | |||
XM_011524883.2 | XP_011523185.1 | 11 | 1408 | |||
XM_011524884.1 | XP_011523186.1 | 11 | 1392 | |||
XM_017024737.1 | XP_016880226.1 | 11 | 1374 | |||
GenBank mRNA | AB209576.1 | - | 10 | 2079 | ||
AF119864.1 | - | 11 | 1253 | |||
AF151827.1 | - | 12 | 1409 | |||
AK026060.1 | - | 12 | 1409 | |||
AL133584.1 | - | 12 | 1409 | |||
BC001398.2 | - | 12 | 1413 | 3051436 | ||
BC009330.2 | - | 12 | 1385 | 4126268 | ||
BC011178.1 | - | 1 | 232 | |||
BC096819.1 | - | 12 | 1394 | 6109001 |