Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024512.4 | NP_078788.2 | 9 | 2023 | ||
XM_011534110.2 | XP_011532412.1 | 9 | 1812 | |||
GenBank mRNA | AJ308569.2 | - | 9 | 1797 | ||
BC029118.1 | - | 9 | 1795 | 5183639 |