Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC26A10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133489.2 | NP_597996.2 | 14 | 2403 | ||
GenBank mRNA | AL050358.1 | - | 9 | 2983 |