Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PTPRF SIRNA Assays ›
Gene Symbol:
PTPRF
Gene Name:
protein tyrosine phosphatase, receptor type F
Gene Aliases:
BNAH2, LAR
Chromosome Location:
Chr.1: 43522238 - 43623672 on Build GRCh38
UniGene ID:
Hs.272062
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
4274
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PTPRF
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
XM_005271079.3
XP_005271136.1
4, 5
601
XM_005271080.3
XP_005271137.1
4, 5
601
XM_005271081.3
XP_005271138.1
4, 5
601
XM_005271082.3
XP_005271139.1
4, 5
601
XM_006710795.3
XP_006710858.1
5, 6
579
XM_006710796.3
XP_006710859.1
4, 5
601
XM_006710797.3
XP_006710860.1
4, 5
601
XM_006710798.3
XP_006710861.1
4, 5
601
XM_006710799.3
XP_006710862.1
4, 5
601
XM_006710800.3
XP_006710863.1
4, 5
601
XM_006710801.3
XP_006710864.1
4, 5
601
XM_011541871.2
XP_011540173.1
5, 6
613
XM_011541872.2
XP_011540174.1
5, 6
599
XM_011541873.2
XP_011540175.1
5, 6
533
XM_017001942.1
XP_016857431.1
2, 3
230
XM_017001943.1
XP_016857432.1
4, 5
601
XM_017001944.1
XP_016857433.1
4, 5
601
XM_017001945.1
XP_016857434.1
4, 5
601
XM_017001946.1
XP_016857435.1
4, 5
601
XM_017001947.1
XP_016857436.1
4, 5
601
XM_017001948.1
XP_016857437.1
4, 5
612
XM_017001949.1
XP_016857438.1
5, 6
577
XM_017001950.1
XP_016857439.1
4, 5
609
GenBank mRNA
AB177856.1
-
4, 5
587
AB177857.1
-
4, 5
560
AB621807.1
-
4, 5
607
BC048768.1
-
4, 5
571
5502598
DA339772.1
-
5, 6
579
DQ234351.1
-
2, 3
245
Y00815.1
-
4, 5
585
Back To Top
More Information
Panther Classification:
Molecular Function -
protein phosphatase
protein modifying enzyme
Gene Ontology Categories:
Function(s)
Process(es)
cell adhesion
transmembrane receptor protein tyrosine phosphatase signaling pathway
chemical synaptic transmission
cell migration
neuron projection regeneration
peptidyl-tyrosine dephosphorylation
regulation of axon regeneration
negative regulation of receptor binding
protein tyrosine phosphatase activity
transmembrane receptor protein tyrosine phosphatase activity
heparin binding
protein complex binding
chondroitin sulfate proteoglycan binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-fwm9x:80/10.239.164.221:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0