Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033504.2 | NP_277039.1 | 988 | |||
GenBank mRNA | BC001418.3 | - | 988 | 3140820 | ||
BC013953.2 | - | 988 | 2987806 | |||
BC023260.1 | - | 866 | 4213210 |