Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: G6PC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138387.3 | NP_612396.1 | 6 | 1570 | ||
NR_028581.1 | 7 | 1764 | ||||
NR_028582.1 | 6 | 1629 | ||||
GenBank mRNA | BC021574.1 | - | 6 | 1550 | 4842511 |