Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERLIN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003790.3 | NP_001003790.1 | 2 | 416 | ||
NM_001003791.2 | NP_001003791.1 | 2 | 187 | |||
NM_007175.6 | NP_009106.1 | 2 | 187 | |||
XM_005273392.2 | XP_005273449.1 | 2 | 347 | |||
XM_006716280.2 | XP_006716343.1 | 2 | 139 | |||
XM_017013000.1 | XP_016868489.1 | 2 | 185 | |||
GenBank mRNA | AK291394.1 | - | 2 | 140 | ||
AK297279.1 | - | 2 | 140 | |||
AL442077.1 | - | 174 | ||||
AY358108.1 | - | 2 | 131 | |||
AY358851.1 | - | 2 | 121 | |||
BC005950.1 | - | 2 | 110 | |||
BC048308.1 | - | 2 | 410 | |||
BC050611.1 | - | 2 | 141 | |||
BC067765.1 | - | 2 | 414 | 5296776 | ||
BI560439.1 | - | 2 | 419 | |||
BP353279.1 | - | 2 | 187 | |||
BP363612.1 | - | 2 | 416 |