Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OR12D3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030959.2 | NP_112221.1 | 1 | 78 | ||
GenBank mRNA | AJ459858.1 | - | 1 | 78 | ||
BC069082.1 | - | 1 | 74 | 7216896 | ||
BC069154.1 | - | 1 | 74 | 7217014 | ||
BC103892.1 | - | 1 | 78 | 40005054 |