Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143830.1 | NP_001137302.1 | 3 | 464 | ||
NM_005256.3 | NP_005247.1 | 2 | 255 | |||
NM_177553.2 | NP_808221.1 | 3 | 406 | |||
XM_011519971.2 | XP_011518273.1 | 3 | 464 | |||
XM_011519972.2 | XP_011518274.1 | 3 | 2459 | |||
XM_011519975.1 | XP_011518277.1 | 3 | 464 | |||
XM_011519978.2 | XP_011518280.1 | 3 | 464 | |||
XM_017017528.1 | XP_016873017.1 | 3 | 1156 | |||
XM_017017529.1 | XP_016873018.1 | 3 | 301 | |||
XM_017017530.1 | XP_016873019.1 | 3 | 494 | |||
XM_017017531.1 | XP_016873020.1 | 3 | 315 | |||
XM_017017532.1 | XP_016873021.1 | 3 | 464 | |||
GenBank mRNA | AK313734.1 | - | 3 | 470 | ||
AU099094.1 | - | 2 | 255 | |||
BC013326.1 | - | 3 | 410 | |||
BC040470.1 | - | 3 | 406 | 4280621 | ||
CR450285.1 | - | 2 | 158 | |||
DA345287.1 | - | 3 | 464 | |||
U95032.1 | - | 2 | 224 |