Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193397.1 | NP_001180326.1 | 2 | 168 | ||
NM_001271832.1 | NP_001258761.1 | 2 | 382 | |||
NM_021131.4 | NP_066954.2 | 3 | 308 | |||
NM_178000.2 | NP_821067.1 | 2 | 382 | |||
NM_178001.2 | NP_821068.1 | 2 | 382 | |||
NM_178003.2 | NP_821070.1 | 2 | 382 | |||
XM_011518834.2 | XP_011517136.1 | 3 | 307 | |||
XM_011518835.2 | XP_011517137.1 | 2 | 360 | |||
XM_011518836.2 | XP_011517138.1 | 3 | 308 | |||
XM_011518837.2 | XP_011517139.1 | 2 | 360 | |||
XM_011518838.2 | XP_011517140.1 | 3 | 289 | |||
XM_017014888.1 | XP_016870377.1 | 2 | 360 | |||
GenBank mRNA | AK222788.1 | - | 2 | 281 | ||
AK297202.1 | - | 2 | 98 | |||
AK302043.1 | - | 2 | 358 | |||
AK302897.1 | - | 2 | 308 | |||
AK307424.1 | - | 2 | 168 | |||
AU133279.1 | - | 2 | 378 | |||
BC002545.2 | - | 3 | 292 | 3138990 | ||
BC011605.2 | - | 3 | 292 | 3140067 | ||
BE732116.1 | - | 2 | 262 | |||
BT020119.1 | - | 2 | 95 | |||
CR749810.1 | - | 2 | 287 | |||
DA052599.1 | - | 2 | 168 | |||
DA387148.1 | - | 2 | 308 | |||
DA737954.1 | - | 2 | 360 | |||
DB009663.1 | - | 3 | 308 | |||
X73478.1 | - | 2 | 284 |