Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHRD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304472.1 | NP_001291401.1 | 23 | 3479 | ||
NM_001304473.1 | NP_001291402.1 | 24 | 3508 | |||
NM_001304474.1 | NP_001291403.1 | 23 | 3453 | |||
NM_003741.3 | NP_003732.2 | 23 | 3482 | |||
NR_130747.1 | 20 | 3536 | ||||
XM_011513254.1 | XP_011511556.1 | 22 | 3071 | |||
XM_011513256.1 | XP_011511558.1 | 21 | 3491 | |||
XM_017007388.1 | XP_016862877.1 | 22 | 3416 | |||
XM_017007389.1 | XP_016862878.1 | 22 | 3413 | |||
XM_017007390.1 | XP_016862879.1 | 21 | 3152 | |||
XM_017007391.1 | XP_016862880.1 | 21 | 3149 | |||
XM_017007392.1 | XP_016862881.1 | 22 | 3068 | |||
XM_017007394.1 | XP_016862883.1 | 21 | 3204 | |||
GenBank mRNA | AB208947.1 | - | 16 | 4454 | ||
AF038198.1 | - | 9 | 1482 | |||
AF076612.1 | - | 20 | 2774 | |||
AF209928.1 | - | 23 | 3482 | |||
AF209930.1 | - | 24 | 3508 | |||
AF283325.1 | - | 23 | 3453 | |||
AK074147.1 | - | 20 | 3289 | |||
AK293456.1 | - | 23 | 3204 | |||
AY358926.1 | - | 23 | 3369 | |||
BC029879.1 | - | 10 | 1612 |