Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLXND1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015103.2 | NP_055918.2 | 36 | 6619 | ||
XM_011512589.1 | XP_011510891.1 | 33 | 6144 | |||
XM_011512592.1 | XP_011510894.1 | 24 | 3702 | |||
GenBank mRNA | AB014520.2 | - | 36 | 6698 | ||
AK026388.1 | - | 12 | 2117 | |||
AY116661.1 | - | 36 | 6699 | |||
BC003526.1 | - | 14 | 2366 | |||
BC011848.2 | - | 13 | 2309 | |||
BC030110.1 | - | 1 | 504 | |||
BC044228.1 | - | 4 | 1344 | |||
BC062465.1 | - | 2 | 1276 | |||
BC150280.1 | - | 36 | 6698 | 9007263 |