Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSTA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319059.1 | NP_001305988.1 | 6 | 765 | ||
NM_145740.4 | NP_665683.1 | 7 | 898 | |||
GenBank mRNA | AK310324.1 | - | 6 | 1773 | ||
BC053578.1 | - | 7 | 733 | 5167338 | ||
BC110891.1 | - | 7 | 735 | 5171697 | ||
BI762344.1 | - | 7 | 777 | |||
M14777.1 | - | 7 | 751 | |||
M15872.1 | - | 7 | 740 | |||
M21758.1 | - | 7 | 751 | |||
M25627.1 | - | 7 | 751 | |||
S49975.1 | - | 6 | 726 |