Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MLKL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152649.3 | NP_689862.1 | 9, 10 | 1946 | ||
XM_005255834.3 | XP_005255891.1 | 10, 11 | 1845 | |||
GenBank mRNA | AK091708.1 | - | 9, 10 | 1689 | ||
AK122998.1 | - | 1, 2 | 3067 |