Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NLRP10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176821.3 | NP_789791.1 | 1 | 229 | ||
XM_011520043.2 | XP_011518345.1 | 2 | 439 | |||
GenBank mRNA | AY154465.1 | - | 1 | 212 | ||
AY489192.1 | - | 1 | 229 | |||
BC104957.1 | - | 1 | 306 | 8143960 |