Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OPCML | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012393.2 | NP_001012393.1 | 4, 5 | 800 | ||
NM_001319103.1 | NP_001306032.1 | 3, 4 | 1078 | |||
NM_001319104.1 | NP_001306033.1 | 3, 4 | 715 | |||
NM_001319105.1 | NP_001306034.1 | 4, 5 | 624 | |||
NM_002545.4 | NP_002536.1 | 3, 4 | 1078 | |||
XM_011542856.2 | XP_011541158.1 | 5, 6 | 1336 | |||
GenBank mRNA | AK289695.1 | - | 3, 4 | 706 | ||
AK299528.1 | - | 5, 6 | 473 | |||
AK299908.1 | - | 4, 5 | 587 | |||
AK315859.1 | - | 3, 4 | 405 | |||
AK316273.1 | - | 3, 4 | 502 | |||
AK316491.1 | - | 4, 5 | 624 | |||
BC074773.2 | - | 3, 4 | 560 | 30915200 | ||
BC117254.1 | - | 3, 4 | 556 | 40125805 | ||
BC126251.1 | - | 3, 4 | 556 | 8991967 | ||
BC143946.1 | - | 3, 4 | 556 | 9052464 | ||
BX537377.1 | - | 4, 5 | 648 | |||
EU562295.1 | - | 3, 4 | 1112 | |||
EU562296.1 | - | 4, 5 | 590 | |||
EU562297.1 | - | 4, 5 | 527 | |||
EU562298.1 | - | 3, 4 | 505 | |||
EU562299.1 | - | 3, 4 | 417 | |||
EU562300.1 | - | 2, 3 | 341 | |||
L34774.1 | - | 3, 4 | 583 |