Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SULT1C2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001056.3 | NP_001047.1 | 7 | 1154 | ||
NM_176825.2 | NP_789795.1 | 8 | 1187 | |||
XM_005264013.3 | XP_005264070.1 | 7 | 975 | |||
GenBank mRNA | AB008164.1 | - | 7 | 824 | ||
AF026303.1 | - | 7 | 786 | |||
AF186251.1 | - | 7 | 1019 | |||
AF186252.1 | - | 7 | 1154 | |||
AF186253.1 | - | 7 | 870 | |||
AF186254.1 | - | 8 | 1052 | |||
AF186255.1 | - | 8 | 1187 | |||
AF186256.1 | - | 8 | 903 | |||
AK297086.1 | - | 7 | 960 | |||
AK298306.1 | - | 7 | 825 | |||
AK313193.1 | - | 7 | 918 | |||
AL832145.1 | - | 3 | 5928 | |||
BC005353.1 | - | 7 | 771 | 4051264 | ||
BT006951.1 | - | 6 | 701 | |||
U66036.1 | - | 7 | 877 |