Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PEPD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000285.3 | NP_000276.2 | 2, 3 | 342 | ||
NM_001166056.1 | NP_001159528.1 | 2, 3 | 342 | |||
GenBank mRNA | AK225037.1 | - | 2, 3 | 223 | ||
AK290756.1 | - | 2, 3 | 223 | |||
AK290781.1 | - | 2, 3 | 238 | |||
AK291561.1 | - | 2, 3 | 240 | |||
AK294619.1 | - | 2, 3 | 240 | |||
BC004305.1 | - | 2, 3 | 213 | 3626134 | ||
BC015027.1 | - | 2, 3 | 243 | 3920275 | ||
BC028295.1 | - | 2, 3 | 214 | 4470066 | ||
BT006692.1 | - | 2, 3 | 209 | |||
CR541669.1 | - | 2, 3 | 209 | |||
DC385464.1 | - | 2, 3 | 270 | |||
DC395475.1 | - | 2, 3 | 344 | |||
J04605.1 | - | 2, 3 | 225 |