Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GOLM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016548.3 | NP_057632.2 | 3 | 406 | ||
NM_177937.2 | NP_808800.1 | 3 | 398 | |||
GenBank mRNA | AF236056.1 | - | 3 | 347 | ||
AK027717.1 | - | 3 | 369 | |||
AK075542.1 | - | 3 | 340 | |||
AK075557.1 | - | 3 | 310 | |||
AK092943.1 | - | 3, 4 | 346 | |||
AY358593.1 | - | 3 | 311 | |||
BC001740.1 | - | 3 | 321 | 3351316 | ||
BQ216766.1 | - | 365 | ||||
CR457201.1 | - | 2 | 197 | |||
DA116177.1 | - | 3 | 399 | |||
EU794616.1 | - | 3 | 365 |