Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ANKRD11 SIRNA Assays ›
Gene Symbol:
ANKRD11
Gene Name:
ankyrin repeat domain 11
Gene Aliases:
ANCO-1, ANCO1, LZ16, T13
Chromosome Location:
Chr.16: 89267619 - 89490561 on Build GRCh38
UniGene ID:
Hs.335003
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
43469
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ANKRD11
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001256182.1
NP_001243111.1
4, 5
612
NM_001256183.1
NP_001243112.1
3, 4
538
NM_013275.5
NP_037407.4
3, 4
541
XM_006721181.1
XP_006721244.1
2, 3
221
XM_011523051.2
XP_011521353.1
4, 5
601
XM_011523053.2
XP_011521355.1
4, 5
707
XM_011523054.2
XP_011521356.1
3, 4
524
XM_011523056.2
XP_011521358.1
3, 4
524
XM_011523057.2
XP_011521359.1
3, 4
524
XM_017023182.1
XP_016878671.1
3, 4
974
XM_017023183.1
XP_016878672.1
4, 5
710
XM_017023184.1
XP_016878673.1
4, 5
707
XM_017023185.1
XP_016878674.1
2, 3
348
XM_017023186.1
XP_016878675.1
6, 7
1084
XM_017023187.1
XP_016878676.1
2, 3
436
GenBank mRNA
AB209186.1
-
2, 3
186
AF121775.1
-
3, 4
298
AK125549.1
-
2, 3
517
AK290994.1
-
2, 3
220
AY373756.1
-
3, 4
541
AY533563.1
-
4, 5
369
BC017437.1
-
3, 4
510
BC025283.1
-
2, 3
165
BC052290.1
-
3, 4
456
BC069013.1
-
3, 4
456
BQ957747.1
-
529
KJ534770.1
-
1, 2
80
KM670019.1
-
139
KM670020.1
-
139
Back To Top
More Information
Gene Ontology Categories:
Function(s)
in utero embryonic development
tissue homeostasis
multicellular organism growth
odontogenesis of dentin-containing tooth
skeletal system morphogenesis
face morphogenesis
bone development
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-567f868b4f-sjrww:80/10.239.163.191:80.
git-commit: 96b0dd45ad55c22ec1de1c352dff25df4a817ab2
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.8.0-2024.05.19-1.0