Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AAMDC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001316957.1 | NP_001303886.1 | 3, 4 | 314 | ||
NM_001316958.1 | NP_001303887.1 | 3, 4 | 314 | |||
NM_001316960.1 | NP_001303889.1 | 3, 4 | 390 | |||
NM_001316961.1 | NP_001303890.1 | 3, 4 | 310 | |||
NM_001316962.1 | NP_001303891.1 | 3, 4 | 314 | |||
NM_024684.3 | NP_078960.1 | 2, 3 | 272 | |||
NR_133637.1 | 2, 3 | 272 | ||||
XM_011544968.2 | XP_011543270.1 | 2, 3 | 272 | |||
XM_011544969.2 | XP_011543271.1 | 2, 3 | 265 | |||
XM_017017620.1 | XP_016873109.1 | 2, 3 | 294 | |||
GenBank mRNA | AF092136.1 | - | 3, 4 | 273 | ||
AK024688.1 | - | 2, 3 | 231 | |||
BC002752.2 | - | 2, 3 | 224 | 3628061 | ||
BC016854.2 | - | 2, 3 | 201 | 3856546 | ||
BI791786.1 | - | 3, 4 | 323 | |||
BM930331.1 | - | 3, 4 | 259 | |||
BP270361.1 | - | 2, 3 | 272 | |||
BX646013.1 | - | 2, 3 | 256 | |||
CN362204.1 | - | 3, 4 | 285 | |||
CR457332.1 | - | 1, 2 | 126 |