Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005880.3 | NP_005871.1 | 1, 2 | 225 | ||
GenBank mRNA | AJ001309.1 | - | 1, 2 | 135 | ||
AK313031.1 | - | 1, 2 | 173 | |||
BC013044.2 | - | 1, 2 | 177 | 3505399 | ||
BC015809.2 | - | 1, 2 | 178 | 3922477 | ||
DB097731.1 | - | 1, 2 | 299 |