Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZC3H7B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017590.5 | NP_060060.3 | 13, 14 | 1743 | ||
GenBank mRNA | AB028954.1 | - | 10, 11 | 1356 | ||
AF188530.1 | - | 13, 14 | 1571 | |||
AK315009.1 | - | 13, 14 | 1685 | |||
BC024313.2 | - | 4, 5 | 410 | |||
BC098378.1 | - | 13, 14 | 1681 | |||
BC152558.1 | - | 13, 14 | 1721 | 8860405 |