Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC38A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001307936.1 | NP_001294865.1 | 2 | 519 | ||
NM_018976.4 | NP_061849.2 | 2 | 519 | |||
GenBank mRNA | AB037803.2 | - | 1 | 79 | ||
AF259799.1 | - | 1 | 99 | |||
AF298897.1 | - | 2 | 430 | |||
AJ344099.1 | - | 2 | 431 | |||
AK001700.1 | - | 2 | 428 | |||
AK291644.1 | - | 2 | 223 | |||
BC040342.1 | - | 2 | 421 | 3874551 | ||
DA725217.1 | - | 2 | 519 |