Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PTPRD SIRNA Assays ›
Gene Symbol:
PTPRD
Gene Name:
protein tyrosine phosphatase, receptor type D
Gene Aliases:
HPTP, HPTPD, HPTPDELTA, PTPD, RPTPDELTA
Chromosome Location:
Chr.9: 8314246 - 10612840 on Build GRCh38
UniGene ID:
Hs.446083
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
4365
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PTPRD
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001040712.2
NP_001035802.1
2
292
NM_001171025.1
NP_001164496.1
2
292
NM_002839.3
NP_002830.1
13
932
NM_130391.3
NP_569075.2
2
292
NM_130392.3
NP_569076.2
2
292
NM_130393.3
NP_569077.2
2
292
XM_006716817.3
XP_006716880.1
12
1230
XM_006716823.2
XP_006716886.1
12
1224
XM_006716825.3
XP_006716888.1
12
1226
XM_006716827.3
XP_006716890.1
12
1223
XM_006716832.3
XP_006716895.1
12
1218
XM_006716833.3
XP_006716896.1
12
1220
XM_006716834.3
XP_006716897.1
12
1221
XM_006716835.3
XP_006716898.1
12
1220
XM_006716837.3
XP_006716900.1
12
1218
XM_006716838.3
XP_006716901.1
12
1220
XM_006716839.3
XP_006716902.1
12
1218
XM_011517992.2
XP_011516294.1
12
1223
XM_017014958.1
XP_016870447.1
12
1229
XM_017014959.1
XP_016870448.1
13
1301
XM_017014960.1
XP_016870449.1
11
1190
XM_017014961.1
XP_016870450.1
11
1174
XM_017014962.1
XP_016870451.1
3
362
XM_017014963.1
XP_016870452.1
12
1229
XM_017014964.1
XP_016870453.1
12
1228
XM_017014965.1
XP_016870454.1
12
1231
XM_017014966.1
XP_016870455.1
12
1229
XM_017014967.1
XP_016870456.1
12
1227
XM_017014968.1
XP_016870457.1
12
1226
XM_017014969.1
XP_016870458.1
12
1229
XM_017014970.1
XP_016870459.1
12
1229
XM_017014971.1
XP_016870460.1
12
1227
XM_017014972.1
XP_016870461.1
12
1229
XM_017014973.1
XP_016870462.1
12
1226
XM_017014974.1
XP_016870463.1
12
1229
XM_017014975.1
XP_016870464.1
12
1224
XM_017014976.1
XP_016870465.1
12
1226
XM_017014977.1
XP_016870466.1
12
1227
XM_017014978.1
XP_016870467.1
12
1226
XM_017014979.1
XP_016870468.1
12
1228
XM_017014980.1
XP_016870469.1
12
1223
XM_017014981.1
XP_016870470.1
12
1218
XM_017014982.1
XP_016870471.1
12
1225
XM_017014983.1
XP_016870472.1
12
1225
XM_017014984.1
XP_016870473.1
12
1225
XM_017014985.1
XP_016870474.1
12
1222
XM_017014986.1
XP_016870475.1
12
1221
XM_017014987.1
XP_016870476.1
12
1221
XM_017014988.1
XP_016870477.1
12
1221
XM_017014989.1
XP_016870478.1
12
1221
XM_017014990.1
XP_016870479.1
12
1220
XM_017014991.1
XP_016870480.1
12
1220
XM_017014992.1
XP_016870481.1
12
1220
XM_017014993.1
XP_016870482.1
12
1218
XM_017014994.1
XP_016870483.1
12
1218
XM_017014995.1
XP_016870484.1
12
1224
GenBank mRNA
AB208869.1
-
1
118
AB211400.1
-
12
900
BC028038.1
-
1004
BC106713.1
-
2
240
40027581
BC106714.1
-
2
240
40027582
BC106715.2
-
2
240
40027584
BC106716.1
-
2
240
40027585
DR001889.1
-
4
367
L38929.1
-
3
342
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
protein dephosphorylation
phosphate-containing compound metabolic process
heterophilic cell-cell adhesion via plasma membrane cell adhesion molecules
transmembrane receptor protein tyrosine phosphatase signaling pathway
chemical synaptic transmission
neuron differentiation
peptidyl-tyrosine dephosphorylation
positive regulation of dendrite morphogenesis
presynaptic membrane assembly
transmembrane receptor protein tyrosine phosphatase activity
receptor binding
protein binding
cell adhesion molecule binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wjc8w:80/10.239.165.44:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0