Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MIB2 SIRNA Assays ›
Gene Symbol:
MIB2
Gene Name:
mindbomb E3 ubiquitin protein ligase 2
Gene Aliases:
ZZANK1, ZZZ5
Chromosome Location:
Chr.1: 1615188 - 1630610 on Build GRCh38
UniGene ID:
Hs.135805
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
43731
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MIB2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001170686.1
NP_001164157.1
6
1052
NM_001170687.1
NP_001164158.1
6
1010
NM_080875.2
NP_543151.2
6
1052
NR_033183.1
6
835
XM_006710372.1
XP_006710435.1
6
1024
XM_011540731.2
XP_011539033.1
6
1726
XM_011540736.2
XP_011539038.1
6
1726
XM_011540737.2
XP_011539039.1
6
1726
XM_011540741.2
XP_011539043.1
6
1726
XM_011540742.2
XP_011539044.1
6
1726
XM_017000349.1
XP_016855838.1
6
1726
XM_017000350.1
XP_016855839.1
6
1726
XM_017000351.1
XP_016855840.1
6
1708
XM_017000352.1
XP_016855841.1
6
1684
XM_017000353.1
XP_016855842.1
6
1726
XM_017000354.1
XP_016855843.1
6
1726
XM_017000355.1
XP_016855844.1
6
1726
XM_017000356.1
XP_016855845.1
6
1726
XM_017000358.1
XP_016855847.1
6
1024
XM_017000359.1
XP_016855848.1
6
1024
XM_017000360.1
XP_016855849.1
6
971
XM_017000361.1
XP_016855850.1
6
1024
XM_017000362.1
XP_016855851.1
6
1064
XM_017000363.1
XP_016855852.1
6
1027
XM_017000365.1
XP_016855854.1
6
754
XM_017000366.1
XP_016855855.1
6
754
GenBank mRNA
AB074480.1
-
6
963
AB076691.1
-
6
921
AB076693.1
-
6
963
AB097000.1
-
6
886
AK097106.1
-
3
387
AK098785.1
-
6
921
AK122751.1
-
6
754
AK122863.1
-
6
953
AK128553.1
-
6
970
AK293642.1
-
5
1032
AK302757.1
-
6
1004
AK309362.1
-
5
1030
AL834527.1
-
1
84
BC016490.1
-
6
889
BC037542.1
-
6
932
5168033
Back To Top
More Information
Panther Classification:
Molecular Function -
ubiquitin-protein ligase
Gene Ontology Categories:
Function(s)
Process(es)
protein polyubiquitination
Notch signaling pathway
positive regulation of I-kappaB kinase/NF-kappaB signaling
actin binding
ubiquitin-protein transferase activity
signal transducer activity
protein binding
zinc ion binding
ligase activity
ubiquitin protein ligase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-jmrqp:80/10.239.165.107:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0