Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IQCE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287499.1 | NP_001274428.1 | 5 | 574 | ||
NM_001287500.1 | NP_001274429.1 | 4 | 526 | |||
NM_001287501.1 | NP_001274430.1 | 3 | 480 | |||
NM_001287502.1 | NP_001274431.1 | 3 | 480 | |||
NM_152558.4 | NP_689771.3 | 5 | 574 | |||
XM_006715676.2 | XP_006715739.1 | 3 | 328 | |||
XM_011515243.1 | XP_011513545.1 | 3 | 455 | |||
XM_017011902.1 | XP_016867391.1 | 5 | 636 | |||
XM_017011903.1 | XP_016867392.1 | 5 | 514 | |||
GenBank mRNA | AB028946.2 | - | 3 | 277 | ||
AK092404.1 | - | 5 | 468 | |||
AK293372.1 | - | 4 | 349 | |||
AK302053.1 | - | 3 | 428 | |||
AK309646.1 | - | 5 | 498 | |||
AL136792.1 | - | 4 | 255 | |||
BC043150.1 | - | 3 | 400 | 5297871 | ||
BC061518.1 | - | 4 | 410 | |||
BC071858.1 | - | 5 | 548 | 5478116 | ||
KJ534872.1 | - | 5 | 364 | |||
KJ535081.1 | - | 3 | 270 |