Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNK17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135111.1 | NP_001128583.1 | 6 | 1139 | ||
NM_031460.3 | NP_113648.2 | 5 | 987 | |||
GenBank mRNA | AF339912.1 | - | 5 | 952 | ||
AF358910.1 | - | 5 | 847 | |||
AK315273.1 | - | 5 | 978 | |||
AY358853.1 | - | 5 | 938 | |||
BC025726.1 | - | 5 | 934 | 5201326 | ||
EU978944.1 | - | 5 | 894 |