Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STT3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278503.1 | NP_001265432.1 | 4, 5 | 396 | ||
NM_001278504.1 | NP_001265433.1 | 2, 3 | 185 | |||
NM_152713.4 | NP_689926.1 | 3, 4 | 308 | |||
XM_011542807.2 | XP_011541109.1 | 3, 4 | 335 | |||
GenBank mRNA | AK290040.1 | - | 3, 4 | 258 | ||
AK290657.1 | - | 3, 4 | 256 | |||
AK295892.1 | - | 2, 3 | 113 | |||
AK308902.1 | - | 3, 4 | 251 | |||
BC020965.2 | - | 3, 4 | 241 | 3891543 | ||
BC048348.1 | - | 2, 3 | 194 | 5561615 | ||
BT007100.1 | - | 2, 3 | 149 | |||
BX649102.1 | - | 4, 5 | 334 | |||
DA577274.1 | - | 3, 4 | 308 | |||
L38961.1 | - | 3, 4 | 255 | |||
L47337.1 | - | 3, 4 | 265 |