Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DYRK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003845.2 | NP_003836.1 | 3 | 243 | ||
NR_104115.1 | 3 | 459 | ||||
GenBank mRNA | AF263541.1 | - | 2 | 143 | ||
AK292322.1 | - | 3 | 237 | |||
AK308260.1 | - | 5 | 464 | |||
AK311576.1 | - | 3 | 424 | |||
BC031244.1 | - | 3 | 237 | 5266287 | ||
DB082653.1 | - | 3 | 243 | |||
GU139743.1 | - | 2 | 213 | |||
GU139746.1 | - | 3 | 432 |