Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MORC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320446.1 | NP_001307375.1 | 3 | 262 | ||
NM_015358.2 | NP_056173.1 | 2 | 176 | |||
XM_011529518.2 | XP_011527820.1 | 2 | 396 | |||
XM_017028311.1 | XP_016883800.1 | 2 | 198 | |||
XM_017028312.1 | XP_016883801.1 | 2 | 119 | |||
GenBank mRNA | AK292957.1 | - | 2 | 127 | ||
BC007311.1 | - | 2 | 135 | |||
BC058002.1 | - | 2 | 180 | |||
BC094779.1 | - | 2 | 133 | 30527767 | ||
BC132731.1 | - | 2 | 130 | 40146753 | ||
BC144116.1 | - | 3 | 216 | |||
BU657895.1 | - | 2 | 167 | |||
D50926.2 | - | 2 | 135 | |||
DA434361.1 | - | 2 | 176 | |||
DA702537.1 | - | 2 | 303 |