Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HOXB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002144.3 | NP_002135.2 | 1 | 566 | ||
GenBank mRNA | BC096191.1 | - | 1 | 566 | 40004081 | |
BC096192.1 | - | 1 | 575 | 40004085 | ||
BC096193.3 | - | 1 | 575 | 40004086 | ||
BC099633.1 | - | 1 | 566 | 40004082 | ||
X16666.1 | - | 1 | 566 |