Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HOXB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002146.4 | NP_002137.4 | 4 | 2379 | ||
XM_005257277.3 | XP_005257334.1 | 5 | 2093 | |||
XM_005257280.3 | XP_005257337.1 | 5 | 2004 | |||
XM_005257282.3 | XP_005257339.1 | 5 | 1569 | |||
XM_006721854.2 | XP_006721917.1 | 5 | 2111 | |||
XM_011524707.2 | XP_011523009.1 | 5 | 11919 | |||
XM_011524708.2 | XP_011523010.1 | 4 | 11499 | |||
XM_011524710.1 | XP_011523012.1 | 5 | 4319 | |||
XM_011524719.1 | XP_011523021.1 | 4 | 6800 | |||
XM_011524720.2 | XP_011523022.1 | 4 | 9491 | |||
XM_011524721.2 | XP_011523023.1 | 4 | 4623 | |||
XM_011524726.2 | XP_011523028.1 | 4 | 1577 | |||
XM_017024557.1 | XP_016880046.1 | 5 | 11667 | |||
XM_017024558.1 | XP_016880047.1 | 5 | 11541 | |||
XM_017024559.1 | XP_016880048.1 | 5 | 11383 | |||
XM_017024560.1 | XP_016880049.1 | 5 | 11627 | |||
XM_017024561.1 | XP_016880050.1 | 5 | 15464 | |||
XM_017024562.1 | XP_016880051.1 | 5 | 11405 | |||
XM_017024563.1 | XP_016880052.1 | 6 | 14060 | |||
GenBank mRNA | AK027609.1 | - | 4 | 2417 | ||
AK291182.1 | - | 5 | 2090 | |||
AK316245.1 | - | 4 | 1612 | |||
U59298.1 | - | 2 | 2970 | |||
X16667.1 | - | 4 | 1884 |