Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UGT3A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_174914.3 | NP_777574.2 | 2, 3 | 321 | ||
NR_031764.1 | 2, 3 | 321 | ||||
XM_017009150.1 | XP_016864639.1 | 2, 3 | 372 | |||
GenBank mRNA | AK075383.1 | - | 2, 3 | 282 | ||
AK303154.1 | - | 2, 3 | 282 | |||
AY358416.1 | - | 2, 3 | 256 | |||
BC103924.1 | - | 2, 3 | 270 | 40007344 | ||
BC103925.1 | - | 2, 3 | 270 | 40007348 | ||
DB110369.1 | - | 2, 3 | 322 |