Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SOX5 SIRNA Assays ›
Gene Symbol:
SOX5
Gene Name:
SRY-box 5
Gene Aliases:
L-SOX5, L-SOX5B, L-SOX5F, LAMSHF
Chromosome Location:
Chr.12: 23529499 - 24562701 on Build GRCh38
UniGene ID:
Hs.657542
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
43936
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SOX5
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001261414.1
NP_001248343.1
6
522
NM_001261415.1
NP_001248344.1
2
238
NM_006940.4
NP_008871.3
2
214
NM_152989.3
NP_694534.1
5
447
XM_011520831.2
XP_011519133.1
2
181
XM_011520832.2
XP_011519134.1
2
185
XM_011520833.2
XP_011519135.1
2
514
XM_011520834.2
XP_011519136.1
6
647
XM_011520835.2
XP_011519137.1
5
699
XM_011520837.2
XP_011519139.1
2
256
XM_011520838.2
XP_011519140.1
2
181
XM_017019888.1
XP_016875377.1
3
633
XM_017019889.1
XP_016875378.1
3
632
XM_017019890.1
XP_016875379.1
6
663
XM_017019891.1
XP_016875380.1
6
521
XM_017019892.1
XP_016875381.1
7
742
XM_017019893.1
XP_016875382.1
4
602
XM_017019894.1
XP_016875383.1
2
2532
XM_017019895.1
XP_016875384.1
2
99
XM_017019896.1
XP_016875385.1
6
663
XM_017019897.1
XP_016875386.1
6
663
XM_017019898.1
XP_016875387.1
6
663
XM_017019899.1
XP_016875388.1
5
375
XM_017019900.1
XP_016875389.1
2
989
XM_017019901.1
XP_016875390.1
2
5445
XM_017019902.1
XP_016875391.1
2
158
XM_017019903.1
XP_016875392.1
2
158
GenBank mRNA
AB081588.1
-
5
445
AB081589.1
-
2
214
AK074317.1
-
2
141
AK096569.1
-
2
613
AK299607.1
-
2
126
AK303970.1
-
2
238
BC060773.1
-
6
523
30343519
JX570584.1
-
3
259
JX570585.1
-
3
259
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
positive regulation of chondrocyte differentiation
asymmetric neuroblast division
positive regulation of cartilage development
cellular response to transforming growth factor beta stimulus
positive regulation of mesenchymal stem cell differentiation
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-nnwcq:80/10.239.162.212:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0