Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIF16B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199865.1 | NP_001186794.1 | 2 | 248 | ||
NM_001199866.1 | NP_001186795.1 | 2 | 248 | |||
NM_024704.4 | NP_078980.3 | 2 | 248 | |||
XM_005260750.3 | XP_005260807.1 | 2 | 227 | |||
XM_005260751.4 | XP_005260808.1 | 2 | 243 | |||
XM_005260752.3 | XP_005260809.1 | 2 | 227 | |||
XM_005260753.3 | XP_005260810.1 | 2 | 225 | |||
XM_005260754.3 | XP_005260811.1 | 2 | 225 | |||
XM_005260755.3 | XP_005260812.1 | 2 | 271 | |||
XM_006723588.3 | XP_006723651.1 | 2 | 225 | |||
XM_017027926.1 | XP_016883415.1 | 2 | 243 | |||
GenBank mRNA | AB046810.2 | - | 2 | 165 | ||
AK095322.1 | - | 2 | 185 | |||
AK307399.1 | - | 2 | 131 | |||
AY166853.1 | - | 2 | 247 | |||
BC050577.1 | - | 2 | 248 | |||
BC150261.1 | - | 2 | 165 | 9007204 | ||
BQ067573.1 | - | 2 | 247 | |||
BX647572.1 | - | 2 | 167 |